Product Details

SNP ID
rs8114297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:36998861 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGCAGGGGATTCTGCATCACTATC[C/G]ATGGCTATTACTCGCTTCTTGGTTC
Phenotype
MIM: 116957
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RBL1 PubMed Links

Gene Details

Gene
RBL1
Gene Name
RB transcriptional corepressor like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323281.1 3003 Missense Mutation ATC,ATG I608M NP_001310210.1
NM_001323282.1 3003 Missense Mutation ATC,ATG I608M NP_001310211.1
NM_002895.4 3003 Missense Mutation ATC,ATG I1035M NP_002886.2
NM_183404.3 3003 Intron NP_899662.1
XM_006723842.2 3003 Missense Mutation ATC,ATG I863M XP_006723905.1
XM_011528955.2 3003 Missense Mutation ATC,ATG I980M XP_011527257.1
XM_011528958.2 3003 Intron XP_011527260.1
XM_017027992.1 3003 Missense Mutation ATC,ATG I931M XP_016883481.1

View Full Product Details