Product Details
- SNP ID
-
rs7644164
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:186668659 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTATACTTCCAGGCATGCGTAGGTG[A/T]TGATGCCTCCGCTAAACCACCACTG
- Phenotype
-
MIM: 142640
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HRG
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116801720] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HRG
- Gene Name
- histidine rich glycoprotein
View Full Product Details