Product Details
- SNP ID
-
rs8004314
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:56122509 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGAAGCATGTGGCTCTCTTGCTGAC[C/T]AATGGCCCATGGTGGTTTTTGGAAA
- Phenotype
-
MIM: 614798
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PELI2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115621852] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PELI2
- Gene Name
- pellino E3 ubiquitin protein ligase family member 2
View Full Product Details