Product Details
- SNP ID
-
rs7508696
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:23774522 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTGTTTTGCCTGGGCCTGGACCTCA[C/G]AAGCCATTGTGACATATTACTGGGC
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RPSAP58
PubMed Links
Gene Details
- Gene
- RPSAP58
- Gene Name
- ribosomal protein SA pseudogene 58
There are no transcripts associated with this gene.
View Full Product Details