Product Details

SNP ID
rs8068012
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:64324246 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCCACTCAAGACATTGTCAGGAAC[A/G]TCTTAAGACCTCAGGAGACCACTTC
Phenotype
MIM: 173445
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PECAM1 PubMed Links

Gene Details

Gene
PECAM1
Gene Name
platelet and endothelial cell adhesion molecule 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000442.4 Intron NP_000433.4
XM_005276880.1 Intron XP_005276937.1
XM_005276881.1 Intron XP_005276938.1
XM_005276882.1 Intron XP_005276939.1
XM_005276883.2 Intron XP_005276940.1
XM_011524889.2 Intron XP_011523191.1
XM_011524890.1 Intron XP_011523192.1
XM_017024738.1 Intron XP_016880227.1
XM_017024739.1 Intron XP_016880228.1
XM_017024740.1 Intron XP_016880229.1
XM_017024741.1 Intron XP_016880230.1

View Full Product Details