Product Details

SNP ID
rs6709389
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:9426011 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAGTGTGATCATACAATACATACT[C/G]TTTTGTCTCTGGCATCTTTCATTTA
Phenotype
MIM: 606029 MIM: 607153
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CPSF3 PubMed Links

Gene Details

Gene
CPSF3
Gene Name
cleavage and polyadenylation specific factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321833.1 Intron NP_001308762.1
NM_001321834.1 Intron NP_001308763.1
NM_001321835.1 Intron NP_001308764.1
NM_001321836.1 Intron NP_001308765.1
NM_016207.3 Intron NP_057291.1
Gene
ITGB1BP1
Gene Name
integrin subunit beta 1 binding protein 1
There are no transcripts associated with this gene.

View Full Product Details