Product Details

SNP ID
rs7084132
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:50800083 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTGCAAAGAAAGGCATGTCTAAG[G/T]CTTGTGATATAGATTTTAAAATTTT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
A1CF PubMed Links

Gene Details

Gene
A1CF
Gene Name
APOBEC1 complementation factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198818.1 8688 UTR 3 NP_001185747.1
NM_001198819.1 8688 UTR 3 NP_001185748.1
NM_001198820.1 8688 UTR 3 NP_001185749.1
NM_014576.3 8688 UTR 3 NP_055391.2
NM_138932.2 8688 UTR 3 NP_620310.1
NM_138933.2 8688 UTR 3 NP_620311.1
XM_005269718.2 8688 UTR 3 XP_005269775.1
XM_005269720.3 8688 UTR 3 XP_005269777.1
XM_011539729.2 8688 UTR 3 XP_011538031.1
XM_011539730.2 8688 UTR 3 XP_011538032.1
XM_017016160.1 8688 UTR 3 XP_016871649.1

View Full Product Details