Product Details

SNP ID
rs6860493
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:43602611 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCCTGGACATCGTTTCTGAGGAAT[C/T]TAACTCCACCTCTCCTGCCAGAATG
Phenotype
MIM: 607878
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NNT PubMed Links

Gene Details

Gene
NNT
Gene Name
nicotinamide nucleotide transhydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012343.3 38 Intron NP_036475.3
NM_182977.2 38 Intron NP_892022.2
XM_005248274.4 38 UTR 5 XP_005248331.1
XM_005248275.4 38 Intron XP_005248332.1
XM_006714461.3 38 Intron XP_006714524.1
XM_011514001.2 38 Intron XP_011512303.1
XM_011514002.2 38 Intron XP_011512304.1
XM_017009293.1 38 Intron XP_016864782.1
Gene
NNT-AS1
Gene Name
NNT antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details