Product Details

SNP ID
rs7255679
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41557611 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAAAACCATACTTGTCACCCTTC[G/T]AGTCCTGTGTGTCTGGAACTAGAAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CEACAM21 PubMed Links

Gene Details

Gene
CEACAM21
Gene Name
carcinoembryonic antigen related cell adhesion molecule 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098506.3 Intron NP_001091976.3
NM_001288773.2 Intron NP_001275702.2
NM_001290113.1 Intron NP_001277042.1
NM_033543.5 Intron NP_291021.4
XM_005278396.4 Intron XP_005278453.1
XM_005278397.3 Intron XP_005278454.1
XM_011527450.1 Intron XP_011525752.1
XM_011527452.1 Intron XP_011525754.1
XM_017027430.1 Intron XP_016882919.1
XM_017027431.1 Intron XP_016882920.1
XM_017027432.1 Intron XP_016882921.1
XM_017027433.1 Intron XP_016882922.1
XM_017027434.1 Intron XP_016882923.1
XM_017027435.1 Intron XP_016882924.1
XM_017027436.1 Intron XP_016882925.1
XM_017027437.1 Intron XP_016882926.1

View Full Product Details