Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000278.4 | Intron | NP_000269.3 | |||
NM_001304569.1 | Intron | NP_001291498.1 | |||
NM_003987.4 | Intron | NP_003978.3 | |||
NM_003988.4 | Intron | NP_003979.2 | |||
NM_003989.4 | Intron | NP_003980.3 | |||
NM_003990.4 | Intron | NP_003981.3 | |||
XM_017016281.1 | Intron | XP_016871770.1 | |||
XM_017016282.1 | Intron | XP_016871771.1 | |||
XM_017016283.1 | Intron | XP_016871772.1 | |||
XM_017016284.1 | Intron | XP_016871773.1 | |||
XM_017016285.1 | Intron | XP_016871774.1 | |||
XM_017016286.1 | Intron | XP_016871775.1 | |||
XM_017016287.1 | Intron | XP_016871776.1 | |||
XM_017016288.1 | Intron | XP_016871777.1 | |||
XM_017016289.1 | Intron | XP_016871778.1 |