Product Details

SNP ID
rs7922613
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:89302352 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGCAAGCTTCCTAAATTCATCTC[C/T]AGAACTGCACGCCTGGGGAGTTGAG
Phenotype
MIM: 147040
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFIT2 PubMed Links

Gene Details

Gene
IFIT2
Gene Name
interferon induced protein with tetratricopeptide repeats 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001547.4 Intron NP_001538.4
Gene
LOC105378419
Gene Name
uncharacterized LOC105378419
There are no transcripts associated with this gene.

View Full Product Details