Product Details
- SNP ID
-
rs6443238
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:9407371 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAAGTTACTGTAGATGCCACTCTTA[C/T]TTAAACCCTAGGCCATGTGTGGAGT
- Phenotype
-
MIM: 615743
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SETD5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs149504845] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SETD5
- Gene Name
- SET domain containing 5
- Gene
- THUMPD3-AS1
- Gene Name
- THUMPD3 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details