Product Details

SNP ID
rs6783705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:10123099 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGAGTAGAAGATAATGACCATGCT[A/G]AGATACCTTTTCTCTCATATGTTGA
Phenotype
MIM: 611183
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BRK1 PubMed Links

Gene Details

Gene
BRK1
Gene Name
BRICK1, SCAR/WAVE actin nucleating complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018462.4 Intron NP_060932.2

View Full Product Details