Product Details

SNP ID
rs7601872
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218455162 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGTATTTTATTTAAAAAGCAGCT[C/G]ACTCATTTGATCTTAACCCTTTGAA
Phenotype
MIM: 193040
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
USP37 PubMed Links

Gene Details

Gene
USP37
Gene Name
ubiquitin specific peptidase 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020935.2 Intron NP_065986.2
XM_005246720.2 Intron XP_005246777.1
XM_005246721.3 Intron XP_005246778.1
XM_005246722.3 Intron XP_005246779.1
XM_005246724.1 Intron XP_005246781.1
XM_011511538.2 Intron XP_011509840.1
XM_011511541.2 Intron XP_011509843.1
XM_017004577.1 Intron XP_016860066.1
Gene
VIL1
Gene Name
villin 1
There are no transcripts associated with this gene.

View Full Product Details