Product Details

SNP ID
rs7123035
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:68062828 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAAGAGATGATGTATGCTGCGGTG[A/G]AGCCCAGCAGGTAGGCAGGTGTGAC
Phenotype
MIM: 118491 MIM: 604592
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHKA PubMed Links

Gene Details

Gene
CHKA
Gene Name
choline kinase alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277.2 Intron NP_001268.2
NM_212469.1 Intron NP_997634.1
XM_017017147.1 Intron XP_016872636.1
XM_017017148.1 Intron XP_016872637.1
Gene
TCIRG1
Gene Name
T-cell immune regulator 1, ATPase H+ transporting V0 subunit a3
There are no transcripts associated with this gene.

View Full Product Details