Product Details
- SNP ID
-
rs7182670
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:45027835 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGGGGTGGTGGGAAGCTTGTTTTTA[C/T]AATGTAGGGTTCACTCGAATGGTAA
- Phenotype
-
MIM: 182500
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SORD
PubMed Links
Gene Details
- Gene
- SORD
- Gene Name
- sorbitol dehydrogenase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003104.5 |
|
Intron |
|
|
NP_003095.2 |
View Full Product Details