Product Details
- SNP ID
-
rs6795918
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:151435472 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CACATTTTAGGCTGAGATGGGGCTA[A/G]CTGAAAGTCAGTGTGATGAAAACCC
- Phenotype
-
MIM: 611318
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
IGSF10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112829133] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IGSF10
- Gene Name
- immunoglobulin superfamily member 10
- Gene
- MED12L
- Gene Name
- mediator complex subunit 12 like
View Full Product Details