Product Details

SNP ID
rs6795197
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:141328230 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGGTGGTGTTTTTCATACAATTT[A/C]ATGTGCATAATACCCGACATTTTTC
Phenotype
MIM: 612218
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZBTB38 PubMed Links

Gene Details

Gene
ZBTB38
Gene Name
zinc finger and BTB domain containing 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080412.2 Intron NP_001073881.2
XM_005247256.4 Intron XP_005247313.1
XM_005247257.1 Intron XP_005247314.1
XM_005247258.1 Intron XP_005247315.1
XM_005247261.3 Intron XP_005247318.1
XM_011512607.1 Intron XP_011510909.1
XM_011512608.1 Intron XP_011510910.1
XM_011512611.1 Intron XP_011510913.1
XM_017006050.1 Intron XP_016861539.1
XM_017006051.1 Intron XP_016861540.1
XM_017006052.1 Intron XP_016861541.1
XM_017006053.1 Intron XP_016861542.1
XM_017006054.1 Intron XP_016861543.1
XM_017006055.1 Intron XP_016861544.1
XM_017006056.1 Intron XP_016861545.1
XM_017006057.1 Intron XP_016861546.1
XM_017006058.1 Intron XP_016861547.1
XM_017006059.1 Intron XP_016861548.1
XM_017006060.1 Intron XP_016861549.1
XM_017006061.1 Intron XP_016861550.1

View Full Product Details