Product Details

SNP ID
rs6774950
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:101779081 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCGCGTCGCGAGCAGGAGCTTGG[C/T]GGCGTAGAGCGCCAGGGTGCCAACT
Phenotype
MIM: 615864
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEP97 PubMed Links

Gene Details

Gene
CEP97
Gene Name
centrosomal protein 97
There are no transcripts associated with this gene.

Gene
NXPE3
Gene Name
neurexophilin and PC-esterase domain family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134456.1 Intron NP_001127928.1
NM_145037.2 Intron NP_659474.1

View Full Product Details