Product Details

SNP ID
rs7838836
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143272589 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAACTGAGGCAGGGGTACTTCTGCA[C/T]AGAGCCAGCAGGGTGGGGAGCAGGG
Phenotype
MIM: 165280
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLI4 PubMed Links

Gene Details

Gene
GLI4
Gene Name
GLI family zinc finger 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138465.3 Intron NP_612474.1
Gene
MINCR
Gene Name
MYC-induced long noncoding RNA
There are no transcripts associated with this gene.

Gene
ZFP41
Gene Name
ZFP41 zinc finger protein
There are no transcripts associated with this gene.

View Full Product Details