Product Details

SNP ID
hCV29284592
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:7045080 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGCACTGAGCATGTGTTCTGTGC[A/G]GATTCTACAGTGGCGGAGAACTCTG
Phenotype
MIM: 608790
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC96 PubMed Links

Gene Details

Gene
CCDC96
Gene Name
coiled-coil domain containing 96
There are no transcripts associated with this gene.

Gene
LOC100129931
Gene Name
uncharacterized LOC100129931
There are no transcripts associated with this gene.

Gene
TADA2B
Gene Name
transcriptional adaptor 2B
There are no transcripts associated with this gene.

View Full Product Details