Product Details

SNP ID
rs7358234
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:32010479 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTTAAAAACACCTCAGGGATTTCA[G/T]ACCAAAATTGACCTATATATATTTT
Phenotype
MIM: 602809
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KIF5B PubMed Links

Gene Details

Gene
KIF5B
Gene Name
kinesin family member 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004521.2 4026 UTR 3 NP_004512.1
XM_017016224.1 4026 UTR 3 XP_016871713.1

View Full Product Details