Product Details

SNP ID
rs7210007
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:15031015 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACTGTAGAGAGCTTGGATGAATAG[C/T]TTAGTGCAAAGATGAATACTGTAGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDRT7 PubMed Links

Gene Details

Gene
CDRT7
Gene Name
CMT1A duplicated region transcript 7 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details