Product Details

SNP ID
hCV29302727
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:13089516 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAAACCTGAGGAACGCACATCGA[A/C]CAGCACCTGCCTGCTTTGCTAGTGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM234B PubMed Links

Gene Details

Gene
FAM234B
Gene Name
family with sequence similarity 234 member B
There are no transcripts associated with this gene.

Gene
GSG1
Gene Name
germ cell associated 1
There are no transcripts associated with this gene.

View Full Product Details