Product Details

SNP ID
rs6580940
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53268313 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGGAAAACGAAGGTTACATTTTGGA[A/T]CCTCGCGGAGTACTGGTCAGGCGGT
Phenotype
MIM: 604143
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ESPL1 PubMed Links

Gene Details

Gene
ESPL1
Gene Name
extra spindle pole bodies like 1, separase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012291.4 15 UTR 5 NP_036423.4
XM_006719705.3 15 UTR 5 XP_006719768.1
XM_011539024.2 15 Intron XP_011537326.1
XM_011539025.2 15 UTR 5 XP_011537327.1
XM_017020253.1 15 UTR 5 XP_016875742.1

View Full Product Details