Product Details

SNP ID
rs7599341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:60767810 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTAGAGAGGTGAAAGCACATTAGT[C/T]TGGTAACTGAGAGTTCCTTGTTGAC
Phenotype
MIM: 616865
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PAPOLG PubMed Links

Gene Details

Gene
PAPOLG
Gene Name
poly(A) polymerase gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022894.3 Intron NP_075045.2
XM_005264500.3 Intron XP_005264557.1
XM_005264501.1 Intron XP_005264558.1
XM_011533041.1 Intron XP_011531343.1
XM_017004723.1 Intron XP_016860212.1
XM_017004724.1 Intron XP_016860213.1

View Full Product Details