Product Details

SNP ID
rs6925236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:52265972 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCAGGCCTCTAGTTTTGAGTTCTA[C/T]TGATGACCAGCAGTTTGTTTCCCAC
Phenotype
MIM: 602693
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MCM3 PubMed Links

Gene Details

Gene
MCM3
Gene Name
minichromosome maintenance complex component 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270472.1 Intron NP_001257401.1
NM_002388.4 Intron NP_002379.3

View Full Product Details