Product Details

SNP ID
rs7677208
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:176188492 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTTTTATTCAATTATAATGTATA[C/T]ACAAAAAGGTAAACAAACAGTAAGT
Phenotype
MIM: 609879 MIM: 609005
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPATA4 PubMed Links

Gene Details

Gene
SPATA4
Gene Name
spermatogenesis associated 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144644.3 Intron NP_653245.2
Gene
WDR17
Gene Name
WD repeat domain 17
There are no transcripts associated with this gene.

View Full Product Details