Product Details

SNP ID
rs6929124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:159790007 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAGGTTTAGTGTCTTTTCCTTGTC[C/T]CTGCTCGGGGAGCGTGAGGCAGATC
Phenotype
MIM: 611831 MIM: 186980
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRPL18 PubMed Links

Gene Details

Gene
MRPL18
Gene Name
mitochondrial ribosomal protein L18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318817.1 Intron NP_001305746.1
NM_014161.4 Intron NP_054880.2
Gene
PNLDC1
Gene Name
PARN like, ribonuclease domain containing 1
There are no transcripts associated with this gene.

Gene
SNORA20
Gene Name
small nucleolar RNA, H/ACA box 20
There are no transcripts associated with this gene.

Gene
SNORA29
Gene Name
small nucleolar RNA, H/ACA box 29
There are no transcripts associated with this gene.

Gene
TCP1
Gene Name
t-complex 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008897.1 Intron NP_001008897.1
NM_030752.2 Intron NP_110379.2

View Full Product Details