Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001207063.1 | Intron | NP_001193992.1 | |||
NM_001207064.1 | Intron | NP_001193993.1 | |||
NM_001207065.1 | Intron | NP_001193994.1 | |||
NM_001207066.1 | Intron | NP_001193995.1 | |||
NM_001338.4 | Intron | NP_001329.1 | |||
XM_011529476.2 | Intron | XP_011527778.1 | |||
XM_011529477.2 | Intron | XP_011527779.1 | |||
XM_011529478.2 | Intron | XP_011527780.1 | |||
XM_011529479.1 | Intron | XP_011527781.1 |