Product Details

SNP ID
rs6429717
Assay Type
Functionally tested
NCBI dbSNP Submissions
33
Location
Chr.1:15156695 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGTCCTAGAATGTCTCAGGTGG[A/C]GGGGACCTTCAGGTCATCAGAGACA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C1orf195 PubMed Links

Gene Details

Gene
C1orf195
Gene Name
chromosome 1 open reading frame 195
There are no transcripts associated with this gene.

Gene
TMEM51
Gene Name
transmembrane protein 51
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136216.1 Intron NP_001129688.1
NM_001136217.1 Intron NP_001129689.1
NM_001136218.1 Intron NP_001129690.1
NM_001319665.1 Intron NP_001306594.1
NM_018022.2 Intron NP_060492.1
XM_005245919.1 Intron XP_005245976.1
XM_011541676.1 Intron XP_011539978.1
XM_017001590.1 Intron XP_016857079.1
Gene
TMEM51-AS1
Gene Name
TMEM51 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details