Product Details

SNP ID
rs6663667
Assay Type
Functionally tested
NCBI dbSNP Submissions
29
Location
Chr.1:160680141 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAACGTTCTCAGAGGCCAGAATCC[C/G]TGATGACTGGGCATGACAGGGGTCA
Phenotype
MIM: 109530
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CD48 PubMed Links

Gene Details

Gene
CD48
Gene Name
CD48 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256030.1 Intron NP_001242959.1
NM_001778.3 Intron NP_001769.2
XM_005245625.1 Intron XP_005245682.1
XM_011510171.1 Intron XP_011508473.1
XM_017002867.1 Intron XP_016858356.1

View Full Product Details