Product Details
- SNP ID
-
rs7284161
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:30634852 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTAGCCTGGAATCTGGATGGACCT[A/C]TTCTGCCCTGGGGCGCGCCGTTTAC
- Phenotype
-
MIM: 613441
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SLC35E4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114083076] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SLC35E4
- Gene Name
- solute carrier family 35 member E4
- Gene
- TCN2
- Gene Name
- transcobalamin 2
There are no transcripts associated with this gene.
View Full Product Details