Product Details
- SNP ID
-
hCV29350123
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:149151210 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGGAGATGGCTGCCTTTCTCCCTG[A/G]GTCTTTTCACATCACCTTTATCCAT
- Phenotype
-
MIM: 611305
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ABLIM3
PubMed Links
Gene Details
- Gene
- ABLIM3
- Gene Name
- actin binding LIM protein family member 3
There are no transcripts associated with this gene.
View Full Product Details