Product Details
- SNP ID
-
rs7028529
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:76613126 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAGGACTCCTCATGTTTTTCAGCAA[A/G]CCATTTCTTTTATTTCACTGTCTGT
- Phenotype
-
MIM: 610691
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PRUNE2
PubMed Links
Gene Details
- Gene
- PRUNE2
- Gene Name
- prune homolog 2
View Full Product Details