Product Details

SNP ID
rs8008017
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24205859 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATACCCTAGCCTTCAGCAGCTCAAG[A/G]TGTTGGCCTTTGGATAGGAGGCTTC
Phenotype
MIM: 610051 MIM: 610711
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHMP4A PubMed Links

Gene Details

Gene
CHMP4A
Gene Name
charged multivesicular body protein 4A
There are no transcripts associated with this gene.

Gene
MDP1
Gene Name
magnesium dependent phosphatase 1
There are no transcripts associated with this gene.

Gene
NEDD8-MDP1
Gene Name
NEDD8-MDP1 readthrough
There are no transcripts associated with this gene.

Gene
TSSK4
Gene Name
testis specific serine kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184739.1 140 UTR 5 NP_001171668.1
NM_001308067.1 140 Intron NP_001294996.1
NM_174944.3 140 UTR 5 NP_777604.2
XM_011536663.2 140 Intron XP_011534965.1
XM_017021226.1 140 UTR 5 XP_016876715.1

View Full Product Details