Product Details

SNP ID
rs6755497
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:113068830 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAAAATTTAATAGGAGTTCAGAGT[A/G]GAGTTACAATAGTAAGGGAAAGATT
Phenotype
MIM: 615296 MIM: 605507
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL1F10 PubMed Links

Gene Details

Gene
IL1F10
Gene Name
interleukin 1 family member 10 (theta)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032556.5 Intron NP_115945.4
NM_173161.2 Intron NP_775184.1
Gene
IL36RN
Gene Name
interleukin 36 receptor antagonist
There are no transcripts associated with this gene.

View Full Product Details