Product Details

SNP ID
hCV29373107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:33658457 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGCCACATTCCATGGTGTTAGC[A/G]GCCAGACTGGCACTCATACCTTCTC
Phenotype
MIM: 603672 MIM: 612478
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACTL10 PubMed Links

Gene Details

Gene
ACTL10
Gene Name
actin like 10
There are no transcripts associated with this gene.

Gene
C20orf144
Gene Name
chromosome 20 open reading frame 144
There are no transcripts associated with this gene.

Gene
CBFA2T2
Gene Name
CBFA2/RUNX1 translocation partner 2
There are no transcripts associated with this gene.

Gene
NECAB3
Gene Name
N-terminal EF-hand calcium binding protein 3
There are no transcripts associated with this gene.

View Full Product Details