Product Details

SNP ID
rs6993472
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:133455295 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGATGAGGCTGGGTGATGGGATCG[C/T]TTGACGGGCTGGGAGGGAGGACAGG
Phenotype
MIM: 607187
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ST3GAL1 PubMed Links

Gene Details

Gene
ST3GAL1
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003033.3 6780 Intron NP_003024.1
NM_173344.2 6780 Intron NP_775479.1
XM_005251025.4 6780 UTR 3 XP_005251082.1
XM_006716617.1 6780 UTR 3 XP_006716680.1
XM_011517225.1 6780 UTR 3 XP_011515527.1
XM_017013736.1 6780 UTR 3 XP_016869225.1
XM_017013737.1 6780 UTR 3 XP_016869226.1

View Full Product Details