Product Details
- SNP ID
-
rs8128071
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:31875390 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTACAGCCGTTTTGAGTGAGGCACC[G/T]CTTTATTATGGGTATGTGGGGATAG
- Phenotype
-
MIM: 606532
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HUNK
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115930070] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HUNK
- Gene Name
- hormonally up-regulated Neu-associated kinase
View Full Product Details