Product Details

SNP ID
rs5930705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:135427033 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAGGAACCAAGTGAAGGGAATCG[A/G]CCTCTGATGAGAGCCTGCCATGTGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMIM10L2A PubMed Links

Gene Details

Gene
SMIM10L2A
Gene Name
small integral membrane protein 10 like 2A
There are no transcripts associated with this gene.

View Full Product Details