Product Details

SNP ID
rs72551377
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.3:121894526 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
TCCAAGGAAACTCTTTTTTCACCTG[G/T]CTCCATTGAAGAGGTACCACCTCGA
Phenotype
MIM: 607659 MIM: 602339
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EAF2 PubMed Links

Gene Details

Gene
EAF2
Gene Name
ELL associated factor 2
There are no transcripts associated with this gene.

Gene
SLC15A2
Gene Name
solute carrier family 15 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145998.1 203 Missense Mutation GGC,GTC G17V NP_001139470.1
NM_021082.3 203 Missense Mutation GGC,GTC G17V NP_066568.3
XM_005247722.3 203 Missense Mutation GGC,GTC G17V XP_005247779.1
XM_006713736.3 203 Missense Mutation GGC,GTC G17V XP_006713799.1
XM_017007074.1 203 Missense Mutation GGC,GTC G17V XP_016862563.1

View Full Product Details