Product Details

SNP ID
rs6629019
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:35919928 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAGAGACATGCAGCTGCGGGTGAT[C/T]CCGGCTGAGGTGAAGTTCCTGGACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CFAP47 PubMed Links

Gene Details

Gene
CFAP47
Gene Name
cilia and flagella associated protein 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304548.1 188 Silent Mutation ATC,ATT I43I NP_001291477.1
NM_152632.3 188 Silent Mutation ATC,ATT I43I NP_689845.2
XM_017029452.1 188 Silent Mutation ATC,ATT I43I XP_016884941.1
XM_017029453.1 188 Silent Mutation ATC,ATT I43I XP_016884942.1

View Full Product Details