Product Details

SNP ID
rs7538199
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:12746267 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGACAGTGAACTCTTGGGGCCG[C/T]TTTATTCTCTGAGGTTAGCAAGGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf158 PubMed Links

Gene Details

Gene
C1orf158
Gene Name
chromosome 1 open reading frame 158
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152290.3 82 UTR 5 NP_689503.3
XM_011542434.1 82 UTR 5 XP_011540736.1

View Full Product Details