Product Details

SNP ID
rs6068434
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:37989109 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTAGCTATCTTCCCCTGCACGGA[A/G]GTCGTTTTTTATTTCTAAATGTTGG
Phenotype
MIM: 614425
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TTI1 PubMed Links

Gene Details

Gene
TTI1
Gene Name
TELO2 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303457.1 Intron NP_001290386.1
NM_014657.2 Intron NP_055472.1

View Full Product Details