Product Details

SNP ID
rs6088526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:34562528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCACCAGCAGCTCTGATGGCCTCA[G/T]GACAAGTACCTGCCATTCTATCACC
Phenotype
MIM: 601242 MIM: 608528
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MAP1LC3A PubMed Links

Gene Details

Gene
MAP1LC3A
Gene Name
microtubule associated protein 1 light chain 3 alpha
There are no transcripts associated with this gene.

Gene
PIGU
Gene Name
phosphatidylinositol glycan anchor biosynthesis class U
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080476.4 Intron NP_536724.1
XM_011528542.2 Intron XP_011526844.1
XM_017027664.1 Intron XP_016883153.1
XM_017027665.1 Intron XP_016883154.1

View Full Product Details