Product Details
- SNP ID
-
rs9894044
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:41437368 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAAAGGTATAACAAGCATCTCTCTT[C/T]GGGTATCCCTCGCCTTAGCCAGCCC
- Phenotype
-
MIM: 604542
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT38
PubMed Links
Gene Details
- Gene
- KRT38
- Gene Name
- keratin 38
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006771.3 |
1838 |
UTR 3 |
|
|
NP_006762.3 |
View Full Product Details