Product Details

SNP ID
rs5997570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:30098484 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGAAGAGGTCAACTATAGGATTG[C/T]ACTTGGTGACTTTGGAGAGAATAGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HORMAD2 PubMed Links

Gene Details

Gene
HORMAD2
Gene Name
HORMA domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011529914.2 Intron XP_011528216.1
XM_011529917.2 Intron XP_011528219.1
XM_011529918.1 Intron XP_011528220.1
XM_017028621.1 Intron XP_016884110.1
XM_017028622.1 Intron XP_016884111.1
XM_017028624.1 Intron XP_016884113.1
XM_017028625.1 Intron XP_016884114.1
XM_017028626.1 Intron XP_016884115.1

View Full Product Details