Product Details

SNP ID
rs6038998
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:872332 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCCTGAGCTCCAAACATTGGAGC[C/T]TAGAAGTGGTGAAGTGGTAAAGCCT
Phenotype
MIM: 603705
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANGPT4 PubMed Links

Gene Details

Gene
ANGPT4
Gene Name
angiopoietin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322809.1 2129 UTR 3 NP_001309738.1
NM_015985.3 2129 UTR 3 NP_057069.1
XM_011529239.2 2129 UTR 3 XP_011527541.1

View Full Product Details