Product Details

SNP ID
rs4653634
Assay Type
Functionally tested
NCBI dbSNP Submissions
43
Location
Chr.1:225403733 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCCTCATAGTTAATACACATCTCT[C/T]CATTTCTCGCCCTTCTTCCCCCAGC
Phenotype
MIM: 603341 MIM: 600024
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAH14 PubMed Links

Gene Details

Gene
DNAH14
Gene Name
dynein axonemal heavy chain 14
There are no transcripts associated with this gene.

Gene
LBR
Gene Name
lamin B receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002296.3 Intron NP_002287.2
NM_194442.2 Intron NP_919424.1
XM_005273125.2 Intron XP_005273182.1
XM_011544185.2 Intron XP_011542487.1

View Full Product Details